missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine 96.0+%, TCI America™

Supplier:  TCI America D000225ML

 View more versions of this product

Catalog No. D000225ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,N-Dimethyldodecylamine
-20°C
149°C
MFCD00008970
2735
YWFWDNVOPHGWMX-UHFFFAOYSA-N
N,N-dimethyldodecan-1-amine
8168
≥96.0% (GC,T)
112-18-5
Yellow
C14H31N
25 mL
n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
CCCCCCCCCCCCN(C)C
213.409
213.41
Liquid

Chemical Identifiers

112-18-5
213.409
YWFWDNVOPHGWMX-UHFFFAOYSA-N
8168
CCCCCCCCCCCCN(C)C
C14H31N
MFCD00008970
n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
N,N-dimethyldodecan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0176

RTECSNumber : JR6600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions