missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylcyclohexylamine 98.0+%, TCI America™
Supplier: TCI America D0705500ML
Specifications
| N,N-Dimethylcyclohexylamine | |
| Yellow | |
| 160°C | |
| MFCD00003844 | |
| 2264 | |
| SVYKKECYCPFKGB-UHFFFAOYSA-N | |
| N,N-dimethylcyclohexanamine | |
| 7415 | |
| 127.23 | |
| Liquid |
| 98-94-2 | |
| 12 (5g/L H2O soln. 20°C) | |
| C8H17N | |
| 500 mL | |
| n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl | |
| CN(C)C1CCCCC1 | |
| 127.231 | |
| CHEBI:59022 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 98-94-2 | |
| 127.231 | |
| SVYKKECYCPFKGB-UHFFFAOYSA-N | |
| 7415 | |
| N,N-dimethylcyclohexanamine |
| C8H17N | |
| MFCD00003844 | |
| n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl | |
| CHEBI:59022 | |
| CN(C)C1CCCCC1 |
Safety and Handling
EINECSNumber : (3)-2274
RTECSNumber : GX1198000
TSCA : Yes