missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylcyclohexylamine 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 408255000

 View more versions of this product

Catalog No. AC408255000


 

Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear
100.0
98.5
Authentic
99%
MFCD00003844
N, N-Dimethylcyclohexylamine
127.23
500g
0.8400g/mL
1.4525 to 1.4545 (20°C, 589nm)
0.25% max.
Glass bottle
C8H17N
98-94-2
99%

Chemical Identifiers

98-94-2
100.0
C8H17N
MFCD00003844
≥98.5%
98.5
99%
127.23
N, N-Dimethylcyclohexylamine, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. Toxic in contact with skin. Toxic if inhaled. Toxic if swallowed. Flammable liquid and vapor. Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 202-715-5

RTECSNumber : GX1198000

TSCA : TSCA