Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethylcyanoacetamide, 98%

$105.58 - $1,476.18

Chemical Identifiers

Chemical Name or Material N,N-Dimethylcyanoacetamide
CAS 7391-40-4
Molecular Formula NCCH2CON(CH3)2
Formula Weight 112.13
MDL Number MFCD00051623
Beilstein 773950
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1098106

 
alfa aesar™
A1098106
5g Each for $105.58

AAA1098122

 
alfa aesar™
A1098122
100g Each for $1,476.18

AAA1098114

 
alfa aesar™
A1098114
25g Each for $435.62
Description & Specifications

Specifications

Formula Weight 112.13
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N,N-Dimethylcyanoacetamide
Melting Point 59° to 66°C
Assay 98%
Quantity 5g
Molecular Formula NCCH2CON(CH3)2
MDL Number MFCD00051623
CAS 7391-40-4
Beilstein 773950
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.