missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylbenzylamine, 99%, ACROS Organics™

Manufacturer:  Acros Organics 159755000

 View more versions of this product

Catalog No. AC159755000



Specifications

Specifications

N, N-Dimethylbenzylamine
54°C
Glass bottle
0.5% max. (K.F.)
Colorless to Yellow
500mL
103-83-3
100.0
C9H13N
MFCD00008329
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
CN(C)CC1=CC=CC=C1
135.21
135.23
99%
0.9000g/mL
Authentic
1.5000 to 1.5020
183.0°C to 184.0°C
-75.0°C
99%
98.5
99%
C6H5CH2N(CH3)2
12, 1019
XXBDWLFCJWSEKW-UHFFFAOYSA-N
N,N-dimethyl-1-phenylmethanamine
7681
Liquid

Chemical Identifiers

103-83-3
135.21
XXBDWLFCJWSEKW-UHFFFAOYSA-N
7681
CN(C)CC1=CC=CC=C1
C9H13N
MFCD00008329
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
N,N-dimethyl-1-phenylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-149-1

RTECSNumber : DP4500000

TSCA : TSCA

SDS Documents