missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylaniline, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A11916AP

 View more versions of this product

Catalog No. AAA11916AP



Description

Description

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
Specifications

Specifications

N, N-Dimethylaniline
500mL
99%
MFCD00008304
507140
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
CN(C)C1=CC=CC=C1
121.183
CHEBI:16269
Liquid
0.956
121-69-7
C8H11N
UN2253
14,3234
JLTDJTHDQAWBAV-UHFFFAOYSA-N
N,N-dimethylaniline
949
121.18
99%

Chemical Identifiers

121-69-7
121.183
JLTDJTHDQAWBAV-UHFFFAOYSA-N
949
N,N-dimethylaniline
C8H11N
MFCD00008304
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
CHEBI:16269
CN(C)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H351-H227

GHS P Statement
P210-P201-P261-P280-P281-P301+P310-P361-P304+P340-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLANILINE

RTECSNumber : BX4725000

TSCA : TSCA

SDS Documents