Learn More
N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 115920010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Dimethylaniline | |
98.5 | |
1.5°C to 2.5°C | |
0.9500g/mL | |
63°C | |
98.5% min. (GC) | |
C8H11N | |
C6H5N(CH3)2 | |
1 L | |
0.95 | |
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene | |
JLTDJTHDQAWBAV-UHFFFAOYSA-N | |
N,N-dimethylaniline | |
949 | |
121.18 | |
Liquid |
121-69-7 | |
100.0 | |
Yellow | |
193.0°C to 194.0°C | |
Authentic | |
Glass Bottle | |
1.5570 to 1.5590 | |
MFCD00008304 | |
01,274; 02,147 | |
15, 3256 | |
Solubility in water: 1g/L (20°C). Other solubilities: freely soluble in alcohol, ether, chloroform, acet, carbon tetrachloride | |
CN(C)C1=CC=CC=C1 | |
121.18 | |
CHEBI:16269 | |
99% |
Chemical Identifiers
121-69-7 | |
121.18 | |
JLTDJTHDQAWBAV-UHFFFAOYSA-N | |
949 | |
N,N-dimethylaniline |
C8H11N | |
MFCD00008304 | |
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene | |
CHEBI:16269 | |
CN(C)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.
Toxic if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-493-5
RUO – Research Use Only