Please login to your online account to display your discounted pricing

N,N-Dimethylaminopentamethyldisilane, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  336640010

 View more versions of this product

Catalog No. AC336640010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.42
Quantity 1g
Density 0.75g/mL
MDL Number MFCD01861710
Molecular Formula C7H21NSi2
CAS 26798-98-1
Chemical Name or Material N, N-Dimethylaminopentamethyldisilane
Linear Formula (CH3)3SiSi(CH3)n(CH3)

Specifications

Chemical Name or Material N, N-Dimethylaminopentamethyldisilane
CAS 26798-98-1
Molecular Formula C7H21NSi2
Linear Formula (CH3)3SiSi(CH3)n(CH3)
Formula Weight 175.42
MDL Number MFCD01861710