Please login to your online account to display your discounted pricing

N,N-Dimethylaminopentamethyldisilane, Acros Organics

Chemical Identifiers

Chemical Name or Material N, N-Dimethylaminopentamethyldisilane
CAS 26798-98-1
Molecular Formula C7H21NSi2
Linear Formula (CH3)3SiSi(CH3)n(CH3)
Formula Weight 175.42
MDL Number MFCD01861710
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC336640010

  SDS
thermo fisher scientific
336640010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336640050

  SDS
thermo fisher scientific
336640050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.42
Quantity 1g
Density 0.75g/mL
MDL Number MFCD01861710
Molecular Formula C7H21NSi2
CAS 26798-98-1
Chemical Name or Material N, N-Dimethylaminopentamethyldisilane
Linear Formula (CH3)3SiSi(CH3)n(CH3)