missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacrylamide (stabilized with MEHQ) 99.0+%, TCI America™
Supplier: TCI America D1091500G
Specifications
| N,N-Dimethylacrylamide (stabilized with MEHQ) | |
| Yellow | |
| C5H9NO | |
| 500 g | |
| n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl | |
| CN(C)C(=O)C=C | |
| 99.13 | |
| 99.13 | |
| Liquid |
| 2680-03-7 | |
| 80°C | |
| MFCD00008626 | |
| 2810 | |
| YLGYACDQVQQZSW-UHFFFAOYSA-N | |
| N,N-dimethylprop-2-enamide | |
| 17587 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 2680-03-7 | |
| 99.13 | |
| YLGYACDQVQQZSW-UHFFFAOYSA-N | |
| 17587 | |
| CN(C)C(=O)C=C |
| C5H9NO | |
| MFCD00008626 | |
| n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl | |
| N,N-dimethylprop-2-enamide |
Safety and Handling
EINECSNumber : (2)-1017
RTECSNumber : AU3230000
TSCA : Yes