Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, EMD Millipore

For headspace gas chromatography SupraSolv™

$343.03 - $343.03

Chemical Identifiers

Chemical Name or Material N,N-Dimethylacetamide
CAS 127-19-5
Molecular Formula CH3CON(CH3)2
Formula Weight 87.12
Synonym DMA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1003990500

 
emd millipore corporation
1003990500
Glass Bottle 500mL Each for $343.03
Description & Specifications

Specifications

Boiling Point 165° to 166°C
Melting Point −20°C
Formula Weight 87.12
pH 4 (200g/L at 20°C)
Quantity 500mL
Description SupraSolv™ solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv headspace solvents are tested for all residual solvents defined by the ICH.
Ignition Point 400°C (752°F)
Odor Amine-like
Packaging Glass Bottle
Refractive Index 1.423
Flash Point 64°C (147.2°F)
Physical Form Liquid
Solubility Information Soluble in water at 20°C
Health Hazard 2 H227 Combustible liquid.
Health Hazard 3 H312 + H332 Harmful in contact with skin or if inhaled.
Vapor Pressure 1.76 hPa (20°C)
Chemical Name or Material N,N-Dimethylacetamide
Color Colorless
Molecular Formula CH3CON(CH3)2
CAS 127-19-5
Synonym DMA
Health Hazard 1 H360 May damage fertility or the unborn child.
Density 0.94g/cm3 at 20°C