Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, EMD Millipore


For headspace gas chromatography SupraSolv™

Quantity 500mL
Packaging Glass Bottle

Manufacturer: emd millipore corporation  1003990500

 View more versions of this product

Catalog No. M1003990500

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 165° to 166°C
Formula Weight 87.12
Melting Point −20°C
pH 4 (200g/L at 20°C)
Quantity 500mL
Description SupraSolv™ solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv headspace solvents are tested for all residual solvents defined by the ICH.
Refractive Index 1.423
Physical Form Liquid
Odor Amine-like
Ignition Point 400°C (752°F)
Flash Point 64°C (147.2°F)
Health Hazard 2 H227 Combustible liquid.
Packaging Glass Bottle
Solubility Information Soluble in water at 20°C
Health Hazard 3 H312 + H332 Harmful in contact with skin or if inhaled.
Vapor Pressure 1.76 hPa (20°C)
Health Hazard 1 H360 May damage fertility or the unborn child.
Color Colorless
Synonym DMA
Chemical Name or Material N,N-Dimethylacetamide
CAS 127-19-5
Molecular Formula CH3CON(CH3)2
Density 0.94g/cm3 at 20°C

Specifications

Chemical Name or Material N,N-Dimethylacetamide
CAS 127-19-5
Molecular Formula CH3CON(CH3)2
Formula Weight 87.12
Synonym DMA
Accessories

H360 May damage fertility or the unborn child.

H312 + H332 Harmful in contact with skin or if inhaled.

H227 Combustible liquid.