missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC264130250
Specifications
N,N-Dimethylacetamide dimethyl acetal | |
8°C | |
Glass bottle | |
0.911 | |
110.0°C to 118.0°C | |
25g | |
97.5 | |
97% | |
MFCD00008476 | |
1,1-dimethoxy-ethyl-dimethyl-amine, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxyethyl dimethylamine, dimethylacetamide dimethyl acetal, dimethylacetamide dimethylacetal, ethanamine, 1,1-dimethoxy-n,n-dimethyl, n,n-dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
CC(N(C)C)(OC)OC | |
133.19 | |
133.19 | |
97% |
0.9110g/mL | |
Authentic | |
1.4160 to 1.4180 (20°C, 589nm) | |
sodium carbonate | |
Colorless | |
18871-66-4 | |
100.0 | |
C6H15NO2 | |
04,166; 05,226; 10,154 | |
FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
1,1-dimethoxy-N,N-dimethylethanamine | |
87835 | |
Liquid |
Chemical Identifiers
18871-66-4 | |
133.19 | |
FBZVZUSVGKOWHG-UHFFFAOYSA-N | |
87835 | |
CC(N(C)C)(OC)OC |
C6H15NO2 | |
MFCD00008476 | |
1,1-dimethoxy-ethyl-dimethyl-amine, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxyethyl dimethylamine, dimethylacetamide dimethyl acetal, dimethylacetamide dimethylacetal, ethanamine, 1,1-dimethoxy-n,n-dimethyl, n,n-dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332 | |
1,1-dimethoxy-N,N-dimethylethanamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and eas
GHS Signal Word: Danger
EINECSNumber : 242-641-