Learn More
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 115690050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Dimethylacetamide | |
127-19-5 | |
100.0 | |
164.0°C to 166.0°C | |
70°C | |
Glass Bottle | |
1.4370 to 1.4390 | |
0.937 | |
CH3CON(CH3)2 | |
Vapor Pressure: 1.7mbar at 25°C | |
0.3% max. | |
01,270; 02,144; 04,165 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12 | |
Pure |
-20.0°C | |
98.5 | |
0.9370g/mL | |
5 mL | |
Authentic | |
Liquid | |
99% | |
C4H9NO | |
MFCD00008686 | |
1.02 mPa.s (20°C) | |
04, 59 | |
15, 3248 | |
Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 | |
99% |
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Safety and Handling
GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0
Signal Word: WARNING!
RUO – Research Use Only