missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, 99%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115690010

 View more versions of this product

Catalog No. AC115690010



Description

Description

Specifications

Specifications

N, N-Dimethylacetamide
98.5
Vapor Pressure: 1.7mbar at 25°C
C4H9NO
MFCD00008686
01,270; 02,144; 04,165
acetamide, n,n-dimethyl, acetdimethylamide, acetyldimethylamine, dimethyl acetamide, dimethylacetamide, dimethylacetone amide, dimethylamide acetate, dmac, n,n-dimethyl acetamide, n,n-dimethylethanamide
CC(=O)N(C)C
87.122
CHEBI:84254
Liquid
Pure
Authentic
1.4370 to 1.4390 (20°C, 589nm)
Undesignated
127-19-5
100.0
99%
CH3CON(CH3)2
04, 59
15, 3248
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
87.12
≥98.5%
0.9370g/mL
Glass Bottle
0.3% max.
1L

Chemical Identifiers

127-19-5
87.122
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
N,N-dimethylacetamide
C4H9NO
MFCD00008686
acetamide, n,n-dimethyl, acetdimethylamide, acetyldimethylamine, dimethyl acetamide, dimethylacetamide, dimethylacetone amide, dimethylamide acetate, dmac, n,n-dimethyl acetamide, n,n-dimethylethanamide
CHEBI:84254
CC(=O)N(C)C
Safety and Handling

Safety and Handling

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0

Signal Word: WARNING!

SDS Documents