Learn More
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 390670025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
N, N-Dimethylacetamide | |
127-19-5 | |
Colorless | |
0.003 meq/g max. | |
164.0°C to 166.0°C | |
70°C | |
Glass Bottle | |
1.4370 to 1.4390 | |
0.937 | |
CH3CON(CH3)2 | |
1.02 mPa.s (20°C) | |
15, 3248 | |
Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 | |
≥99.8% |
-20.0°C | |
0.046 at 310nm, 0.301 at 285nm, 0.7 (1 cm cell vs HPLC−grade water) at 275nm | |
0.0005 meq/g max. | |
0.9370g/mL | |
2.5 L | |
Authentic | |
Liquid | |
99.8+% | |
C4H9NO | |
MFCD00008686 | |
0.05% max. | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12 | |
HPLC |
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Safety and Handling
GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only