Learn More
N,N-Dimethylacetamide, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
$122.20 - $327.94
Chemical Identifiers
CAS | 127-19-5 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00008686 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
IUPAC Name | N,N-dimethylacetamide |
SMILES | CN(C)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC375231000
|
Thermo Scientific Chemicals
375231000 |
100 mL | AcroSeal™ Glass Bottle |
N/A
|
|
||||
AC375230010
|
Thermo Scientific Chemicals
375230010 |
1 L | AcroSeal™ Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Specifications
127-19-5 | |
100.0 | |
Colorless | |
164.0°C to 166.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.4370 to 1.4390 | |
MFCD00008686 | |
0.937 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
CHEBI:84254 | |
99.5% | |
Liquid |
99.4 | |
-20.0°C | |
0.9370g/mL | |
70°C | |
99.4% min. (GC) | |
C4H9NO | |
CH3CON(CH3)2 | |
100 mL | |
15, 3248 | |
Solubility in water: soluble. Other solubilities: soluble in acetone,diethyl ether,benzene,,chloroform,ethanol and toluene | |
CN(C)C(C)=O | |
87.12 | |
1.02 mPa.s (20°C) | |
87.12 | |
Water <50ppm | |
N, N-Dimethylacetamide, Over Molecular Sieves |
Safety and Handling
GHS H Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 204-826-4
RUO – Research Use Only