Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, 99.5+%, for analysis, ACROS Organics™

$166.21 - $166.21

Chemical Identifiers

Chemical Name or Material N, N-Dimethylacetamide
Name Note For analysis
CAS 127-19-5
CAS Min % 99.5
CAS Max % 100.0
Purity Grade For analysis
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C4H9NO
Linear Formula CH3CON(CH3)2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390800010

  SDS
fisher scientific acros
AC390800010
Glass bottle 1L Each for $166.21
Description & Specifications

Specifications

Assay ≥99.50%
Physical Form Liquid
Formula Weight 87.12
Quantity 1L
Color Clear
Merck Index 15, 3248
Purity Grade For analysis
Density 0.9370g/mL
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.1ppm max.
Name Note For analysis
Water 0.05% max. (K.F.)
Assay Percent Range 99.5+%
Molecular Formula C4H9NO
Linear Formula CH3CON(CH3)2
MDL Number MFCD00008686
CAS 127-19-5
Cadmium (Cd) 0.05ppm max.
CAS Min % 99.5
Health Hazard 1 Danger
Calcium (Ca) 0.5ppm max.
Chromium (Cr) 0.02ppm max.
Copper (Cu) 0.02ppm max.
Chemical Name or Material N, N-Dimethylacetamide
Magnesium (Mg) 0.1ppm max.
Manganese (Mn) 0.02ppm max.
Nickel (Ni) 0.02ppm max.
Packaging Glass bottle
Residue after Evaporation 0.0005% max.
Sulfate 0.001% max.
Zinc (Zn) 0.1ppm max.
Chloride 0.001% max.
CAS Max % 100.0
Health Hazard 2 GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acidity 0.0005 meq/g max.

For analysis

Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.