missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylacetamide, 98% min., EMD Millipore

Manufacturer:  Millipore Sigma DX154530

 View more versions of this product

Catalog No. MDX154530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N, N-Dimethylacetamide
98% Min.
CH3CON(CH3)2
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
87.12
Colorless
127-19-5
C4H9NO
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CC(=O)N(C)C
87.122
CHEBI:84254
Liquid
200L

Chemical Identifiers

127-19-5
87.122
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CHEBI:84254
CC(=O)N(C)C
C4H9NO
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
N,N-dimethylacetamide
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Documents