Learn More
N,N-Dimethyl-p-toluidine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 116905000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Dimethyl-p-toluidine | |
99-97-8 | |
100.0 | |
0.9300g/mL | |
83°C | |
98.5% min. (GC) | |
C9H13N | |
CH3C6H4N(CH3)2 | |
500 g | |
0.93 | |
Solubility in water: immiscible | |
CN(C)C1=CC=C(C)C=C1 | |
135.21 | |
135.21 | |
Liquid |
99% | |
98.5 | |
Yellow | |
211.0°C | |
Authentic | |
Glass bottle | |
1.5450 to 1.5470 | |
MFCD00008316 | |
12, 902 | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
N,N,4-trimethylaniline | |
7471 | |
99% |
Chemical Identifiers
99-97-8 | |
135.21 | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
7471 | |
CN(C)C1=CC=C(C)C=C1 |
C9H13N | |
MFCD00008316 | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
N,N,4-trimethylaniline |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Call a
GHS Signal Word: Danger
EINECSNumber : 202-805-4
RTECSNumber : XU5803000
TSCA : TSCA
Recommended Storage : product darkens on exposure to air, light and moisture
RUO – Research Use Only