Please login to your online account to display your discounted pricing

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%, ACROS Organics™

$79.49 - $79.49

Chemical Identifiers

Chemical Name or Material N, N-Dimethyl-p-phenylenediamine dihydrochloride
CAS 536-46-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H12N2·2HCl
Linear Formula (CH3)2NC6H4NH2·2HCl
Formula Weight 209.12
MDL Number MFCD00012991
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC204351000

  SDS
thermo fisher scientific
AC204351000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC204350250

  SDS
thermo fisher scientific
204350250
Glass bottle 25g Each for $79.49
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 209.12
Quantity 100g
Color White to beige to gray
Beilstein 13, 73
Fieser 01,293
Merck Index 10, 3248
CAS Max % 100.0
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine dihydrochloride
Assay Percent Range 99%
Molecular Formula C8H12N2·2HCl
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2NC6H4NH2·2HCl
MDL Number MFCD00012991
CAS 536-46-9
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.