Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-p-phenylenediamine, 96%

Chemical Identifiers

Chemical Name or Material N,N-Dimethyl-p-phenylenediamine
CAS 99-98-9
Molecular Formula C8H12N2
Formula Weight 136.2
MDL Number MFCD00007860
UN Number UN2811
Beilstein 508105
Merck Index 14,3254
Synonym 4-Amino-N,N-dimethylaniline; 4-Dimethylaminoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA159620B

 
alfa aesar™
A159620B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1596218

 
alfa aesar™
A1596218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1596230

 
alfa aesar™
A1596230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.2
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine
TSCA TSCA
Melting Point 34° to 38°C
Boiling Point 262°C
Assay 96%
Quantity 1000g
Flash Point 130°C (266°F)
Molecular Formula C8H12N2
MDL Number MFCD00007860
CAS 99-98-9
UN Number UN2811
Beilstein 508105
Merck Index 14,3254
Synonym 4-Amino-N,N-dimethylaniline; 4-Dimethylaminoaniline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number ST0874000

Applications
N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

Solubility
Soluble in Water (11 g/L at20°C), diethyl ether, petroleum ether, alcohol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light and air sensitive. Store under inert gas. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.