missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethyl-p-phenylenediamine, 97%, ACROS Organics™
$28.81 - $879.56
Chemical Identifiers
CAS | 99-98-9 |
---|---|
Molecular Formula | C8H12N2 |
Molecular Weight (g/mol) | 136.2 |
MDL Number | MFCD00007860 |
InChI Key | BZORFPDSXLZWJF-UHFFFAOYSA-N |
Synonym | 4-amino-n,n-dimethylaniline, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, dmpd, n,n-dimethyl-1,4-phenylenediamine, n,n-dimethyl-p-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-amino-n,n-dimethylaniline, p-aminodimethylaniline, p-dimethylamino aniline |
PubChem CID | 7472 |
ChEBI | CHEBI:15783 |
IUPAC Name | 4-N,4-N-dimethylbenzene-1,4-diamine |
SMILES | CN(C)C1=CC=C(C=C1)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC408460050
|
Thermo Fisher Scientific
408460050 |
5g | Glass bottle |
Each for $28.81
|
|
AC408460250
|
Thermo Fisher Scientific
AC408460250 |
25g | Glass bottle |
Each for $94.18
|
|
AC408461000
|
Thermo Fisher Scientific
AC408461000 |
100g | Glass bottle |
Each for $239.07
|
|
AC408465000
|
Thermo Fisher Scientific
AC408465000 |
500g | Glass bottle |
Each for $879.56
|
|
Chemical Identifiers
99-98-9 | |
136.2 | |
BZORFPDSXLZWJF-UHFFFAOYSA-N | |
7472 | |
4-N,4-N-dimethylbenzene-1,4-diamine |
C8H12N2 | |
MFCD00007860 | |
4-amino-n,n-dimethylaniline, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, dmpd, n,n-dimethyl-1,4-phenylenediamine, n,n-dimethyl-p-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-amino-n,n-dimethylaniline, p-aminodimethylaniline, p-dimethylamino aniline | |
CHEBI:15783 | |
CN(C)C1=CC=C(C=C1)N |
Specifications
1.0900g/mL | |
Authentic | |
1.09 | |
Brown | |
5g | |
96.0 | |
96% min. (GC) | |
NH2C6H4N(CH3)2 | |
15, 3278 | |
Solubility in water: 11g/L (20°C). Other solubilities: soluble in alcohol,chloroform,ether,benzene | |
CN(C)C1=CC=C(C=C1)N | |
136.2 | |
CHEBI:15783 | |
Crystalline Low Melting Solid | |
N, N-Dimethyl-p-phenylenediamine, Practical |
130°C | |
Glass bottle | |
262.0°C | |
35.0°C | |
99-98-9 | |
100.0 | |
C8H12N2 | |
MFCD00007860 | |
4-amino-n,n-dimethylaniline, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, dmpd, n,n-dimethyl-1,4-phenylenediamine, n,n-dimethyl-p-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-amino-n,n-dimethylaniline, p-aminodimethylaniline, p-dimethylamino aniline | |
BZORFPDSXLZWJF-UHFFFAOYSA-N | |
4-N,4-N-dimethylbenzene-1,4-diamine | |
7472 | |
136.2 | |
97% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
GHS Signal Word: Danger
EINECSNumber : 202-807-5