Please login to your online account to display your discounted pricing

N,N-Dimethyl-o-toluidine, +99%, ACROS Organics™

$54.59 - $54.59

Chemical Identifiers

Chemical Name or Material N, N-Dimethyl-o-toluidine
CAS 609-72-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00035789
Synonym N, N, 2-Trimethylbenzenamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC408621000

  SDS
thermo fisher scientific
408621000
Glass bottle 100g Each for $54.59
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 135.21
Quantity 100g
Color Clear colorless to amber
Density 0.93g/mL
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material N, N-Dimethyl-o-toluidine
Assay Percent Range 99+%
CAS 609-72-3
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00035789
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Additional Information Light and moisture
Molecular Formula C9H13N
Synonym N, N, 2-Trimethylbenzenamine
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Storage Information product darkens on exposure to air

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.