Please login to your online account to display your discounted pricing

N,N-Dimethyl-o-toluidine, +99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  408621000

 View more versions of this product

Catalog No. AC408621000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 135.21
Quantity 100g
Color Clear colorless to amber
Density 0.93g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
CAS 609-72-3
Chemical Name or Material N, N-Dimethyl-o-toluidine
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00035789
Packaging Glass bottle
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Storage Information product darkens on exposure to air
Additional Information Light and moisture
Synonym N, N, 2-Trimethylbenzenamine
Molecular Formula C9H13N
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.

Specifications

Chemical Name or Material N, N-Dimethyl-o-toluidine
CAS 609-72-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00035789
Synonym N, N, 2-Trimethylbenzenamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.

SDS & Safety Documents