missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6575803
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine | |
Yellow | |
MFCD00014858 | |
3602172 | |
XRTKMBBIDHUQNP-UHFFFAOYSA-N | |
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine | |
67160 | |
90% | |
Liquid |
123-67-1 | |
C13H32N4 | |
1 g | |
6,10-dimethyl-2,6,10,14-tetraazapentadecane, 2,6,10,14-tetraazapentadecane, 6,10-dimethyl, n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine, 1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth, 1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl, ksc496s7p | |
CNCCCN(C)CCCN(C)CCCNC | |
244.427 | |
244.43 | |
Technical |
Chemical Identifiers
123-67-1 | |
244.427 | |
XRTKMBBIDHUQNP-UHFFFAOYSA-N | |
67160 | |
CNCCCN(C)CCCN(C)CCCNC |
C13H32N4 | |
MFCD00014858 | |
6,10-dimethyl-2,6,10,14-tetraazapentadecane, 2,6,10,14-tetraazapentadecane, 6,10-dimethyl, n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine, 1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl, n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth, 1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl, ksc496s7p | |
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-641-9
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only