missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-N'-ethylethylenediamine, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 155530250

 View more versions of this product

Catalog No. AC155530250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

116.21
≥97.5%
Clear colorless to slightly yellow
100.0
Glass bottle
C2H5NHCH2CH2N(CH3)2
MFCD00009034
123-83-1
N, N-Dimethyl-N'-ethylethylenediamine
Liquid
25g
0.7300g/mL
1.4210 to 1.4240 (20°C, 589nm)
97.5
Authentic
98%
98%
C6H16N2

Chemical Identifiers

123-83-1
100.0
C6H16N2
116.21
N, N-Dimethyl-N'-ethylethylenediamine, 98%
97.5
98%
C2H5NHCH2CH2N(CH3)2
MFCD00009034
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-656-0

TSCA : TSCA