missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-N'-ethylethylenediamine, 98%, Acros Organics

Chemical Identifiers

CAS 123-83-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H16N2
Linear Formula C2H5NHCH2CH2N(CH3)2
Formula Weight 116.21
MDL Number MFCD00009034
Chemical Name or Material N, N-Dimethyl-N'-ethylethylenediamine, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC155530250 SDS Thermo Fisher Scientific
155530250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

123-83-1
100.0
C6H16N2
116.21
N, N-Dimethyl-N'-ethylethylenediamine, 98%
97.5
98%
C2H5NHCH2CH2N(CH3)2
MFCD00009034
≥97.5%

Specifications

116.21
25g
0.7300g/mL
1.4210 to 1.4240 (20°C, 589nm)
97.5
Authentic
98%
C6H16N2
≥97.5%
Liquid
Clear colorless to slightly yellow
100.0
Glass bottle
C2H5NHCH2CH2N(CH3)2
MFCD00009034
123-83-1
N, N-Dimethyl-N'-ethylethylenediamine, 98%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-656-0

TSCA : TSCA