missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%

Manufacturer:  Alfa Aesar™ L0040314

 View more versions of this product

Catalog No. AAL0040314



Description

Description

Applications
it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

Solubility
Soluble in water, chloroform and dichloromethane. Slightly soluble in ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

N,N-Dimethyl-m-phenylenediamine dihydrochloride
25g
C8H14Cl2N2
UN2811
1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, 3-amino-n,n-dimethylaniline, acmc-20a5k9, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, pubchem14991
BZJPIQKDEGXVFG-UHFFFAOYSA-N
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
77124
99%
216°C (decomposition)
3575-32-4
MFCD00012971
3694384
Soluble in water,chloroform and dichloromethane. Slightly soluble in ethyl acetate.
CN(C)C1=CC=CC(=C1)N.Cl.Cl
209.114
209.12

Chemical Identifiers

3575-32-4
209.114
BZJPIQKDEGXVFG-UHFFFAOYSA-N
77124
CN(C)C1=CC=CC(=C1)N.Cl.Cl
C8H14Cl2N2
MFCD00012971
1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, 3-amino-n,n-dimethylaniline, acmc-20a5k9, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, pubchem14991
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H315-H317-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 222-693-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents