Please login to your online account to display your discounted pricing

N,N-Dimethyl-4,4'-azodianiline, 95%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  209200500

 View more versions of this product

Catalog No. AC209200500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.31
Physical Form Crystalline Powder
Assay ≥94%
Quantity 50g
Color Dark orange to rust brown
TSCA TSCA
RTECS Number BX5020000
EINECS Number 208-712-5
Beilstein 16, 335
CAS Min % 94.0
CAS Max % 100.0
Synonym 4-Amino-4'-(dimethylamino)azobenzene
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause damage to organs.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Suspected of causing cancer.
Suspected of causing genetic defects.
Chemical Name or Material N, N-Dimethyl-4, 4'-azodianiline
Linear Formula (CH3)2NC6H4N=NC6H4NH2
MDL Number MFCD00010204
Molecular Formula C14H16N4
CAS 539-17-3
Assay Percent Range 95%

Specifications

Chemical Name or Material N, N-Dimethyl-4, 4'-azodianiline
CAS 539-17-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H16N4
Linear Formula (CH3)2NC6H4N=NC6H4NH2
Formula Weight 240.31
MDL Number MFCD00010204
Synonym 4-Amino-4'-(dimethylamino)azobenzene
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause damage to organs.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Suspected of causing cancer.
Suspected of causing genetic defects.

SDS & Safety Documents