missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-4,4'-azodianiline, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 209200100

 View more versions of this product

Catalog No. AC209200100


 

Specifications Description & Specifications

Specifications

≥94%
Crystalline Powder
Dark orange to rust brown
100.0
4-Amino-4'-(dimethylamino)azobenzene
Glass bottle
539-17-3
95%
(CH3)2NC6H4N=NC6H4NH2
240.31
10g
16, 335
94.0
Authentic
C14H16N4
N, N-Dimethyl-4, 4'-azodianiline
MFCD00010204

Chemical Identifiers

539-17-3
100.0
C14H16N4
240.31
4-Amino-4'-(dimethylamino)azobenzene
≥94%
94.0
95%
(CH3)2NC6H4N=NC6H4NH2
MFCD00010204
N, N-Dimethyl-4, 4'-azodianiline

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause damage to organs.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Suspected of causing cancer.
Suspected of causing genetic defects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 208-712-5

RTECSNumber : BX5020000

TSCA : TSCA