missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-4-(2-pyridylazo)aniline, 98%

$55.55 - $233.97

Chemical Identifiers

CAS 13103-75-8
Molecular Formula C13H14N4
Formula Weight 226.28
MDL Number MFCD00010435
Beilstein 185623
Synonym 4-(2-Pyridylazo)-N,N-dimethylaniline
Chemical Name or Material N,N-Dimethyl-4-(2-pyridylazo)aniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2378903 SDS Alfa Aesar™
B2378903
1g Each for $55.55
AAB2378906 SDS Alfa Aesar™
B2378906
5g Each for $233.97
Specifications Description & Specifications

Chemical Identifiers

13103-75-8
226.28
185623
N,N-Dimethyl-4-(2-pyridylazo)aniline
C13H14N4
MFCD00010435
4-(2-Pyridylazo)-N,N-dimethylaniline

Specifications

108° to 111°C
13103-75-8
226.28
185623
N,N-Dimethyl-4-(2-pyridylazo)aniline
1g
C13H14N4
MFCD00010435
4-(2-Pyridylazo)-N,N-dimethylaniline
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning