missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-3-piperidinemethylamine dihydrochloride, 95%

Manufacturer:  Alfa Aesar™ H3345503

 View more versions of this product

Catalog No. AAH3345503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N,N-Dimethyl-3-piperidinemethylamine dihydrochloride
~270°C
1g
MFCD13561484
215.16
215.16
1176419-81-0
95%
C8H20Cl2N2
FGUAPATZXBHZIQ-UHFFFAOYNA-N
46735244

Chemical Identifiers

1176419-81-0
215.16
FGUAPATZXBHZIQ-UHFFFAOYNA-N
C8H20Cl2N2
MFCD13561484
46735244
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents