missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-3-nitroaniline 98%, ACROS Organics™

Chemical Identifiers

CAS 619-31-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H10N2O2
Linear Formula O2NC6H4N(CH3)2
Formula Weight 166.18
MDL Number MFCD00007236
Chemical Name or Material N, N-Dimethyl-3-nitroaniline, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC116470050 SDS Thermo Fisher Scientific
116470050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC116470250 SDS Thermo Fisher Scientific
116470250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

619-31-8
100.0
C8H10N2O2
166.18
N, N-Dimethyl-3-nitroaniline, 98%
97.5
98%
O2NC6H4N(CH3)2
MFCD00007236
≥97.5%

Specifications

Orange to orange-red
5g
97.5
100.0
Crystals or Crystalline Powder
C8H10N2O2
166.18
12, 701
≥97.5%
57° to 61°C
619-31-8
Authentic
Glass bottle
98%
O2NC6H4N(CH3)2
MFCD00007236
N, N-Dimethyl-3-nitroaniline, 98%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-590-3

TSCA : TSCA