missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-3-nitroaniline, 98%

$49.20 - $144.65

Chemical Identifiers

CAS 619-31-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007236
InChI Key CJDICMLSLYHRPT-UHFFFAOYSA-N
Synonym 3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
PubChem CID 69269
IUPAC Name N,N-dimethyl-3-nitroaniline
SMILES CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0040106 View Documents Alfa Aesar™
L0040106
5g Each for $49.20
 
AAL0040114 View Documents Alfa Aesar™
L0040114
25g Each for $144.65
 
Specifications

Chemical Identifiers

619-31-8
166.18
CJDICMLSLYHRPT-UHFFFAOYSA-N
69269
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
C8H10N2O2
MFCD00007236
3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
N,N-dimethyl-3-nitroaniline

Specifications

57°C to 60°C
619-31-8
MFCD00007236
1841745
CJDICMLSLYHRPT-UHFFFAOYSA-N
N,N-dimethyl-3-nitroaniline
69269
98%
5g
C8H10N2O2
UN2811
3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
166.18
166.18
N,N-Dimethyl-3-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA