Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride, 96%

$106.23 - $381.22

Chemical Identifiers

Chemical Name or Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
CAS 902836-08-2
Molecular Formula C12H18N4O·HCl
Formula Weight 270.76
MDL Number MFCD12407084
Synonym N,N-Dimethyl-2-(1-piperazinyl)pyridine-3-carboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5167903

  SDS
alfa aesar™
H5167903
1g Each for $381.22

AAH51679MD

  SDS
alfa aesar™
H51679MD
250mg Each for $106.23
Description & Specifications

Specifications

Formula Weight 270.76
Quantity 1g
Molecular Formula C12H18N4O·HCl
MDL Number MFCD12407084
Synonym N,N-Dimethyl-2-(1-piperazinyl)pyridine-3-carboxamide hydrochloride
Chemical Name or Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
Assay 96%
CAS 902836-08-2