Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dimethyl-1-phenylethylamine, 97%

$40.95 - $578.20

Chemical Identifiers

Chemical Name or Material N,N-Dimethyl-1-phenylethylamine
CAS 2449-49-2
Molecular Formula C10H15N
Formula Weight 149.23g/mol
MDL Number MFCD00064770
Synonym N,N,alpha-Trimethylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1798906

 
alfa aesar™
A17989-06
5g Each for $40.95

AAA1798914

 
alfa aesar™
A17989-14
25g Each for $578.20
Description & Specifications

Specifications

Boiling Point 193°C
Quantity 5g
Chemical Name or Material N,N-Dimethyl-1-phenylethylamine
CAS 2449-49-2
Assay 97%
Molecular Formula C10H15N
Formula Weight 149.23g/mol
MDL Number MFCD00064770
Synonym N,N,alpha-Trimethylbenzylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DP6825000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.