Learn More
N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals
$128.99 - $473.78
Chemical Identifiers
CAS | 111-33-1 |
---|---|
Molecular Formula | C5H16N2 |
Molecular Weight (g/mol) | 104.20 |
MDL Number | MFCD00008292 |
InChI Key | UQUPIHHYKUEXQD-UHFFFAOYSA-P |
Synonym | n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin |
PubChem CID | 66978 |
IUPAC Name | N,N'-dimethylpropane-1,3-diamine |
SMILES | C[NH2+]CCC[NH2+]C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1690106
|
Thermo Scientific Chemicals
A1690106 |
5 g |
Each for $128.99
|
|
|||||
AAA1690114
|
Thermo Scientific Chemicals
A1690114 |
25 g |
Each for $473.78
|
|
|||||
Description
Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.
Solubility
Fully miscible in water, 1e+006 mg/L @ 25°C (est).
Notes
Air sensitive. Store away from oxidizing agents, carbon dioxide and air. Store under inert gas.
Chemical Identifiers
111-33-1 | |
104.20 | |
UQUPIHHYKUEXQD-UHFFFAOYSA-P | |
66978 | |
C[NH2+]CCC[NH2+]C |
C5H16N2 | |
MFCD00008292 | |
n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin | |
N,N'-dimethylpropane-1,3-diamine |
Specifications
111-33-1 | |
143°C to 146°C | |
Amine-like | |
MFCD00008292 | |
UN2733 | |
n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin | |
UQUPIHHYKUEXQD-UHFFFAOYSA-P | |
N,N'-dimethylpropane-1,3-diamine | |
66978 | |
97% |
0.817 | |
32°C (90°F) | |
C5H16N2 | |
5 g | |
Air sensitive; Hygroscopic | |
Fully miscible in water,1e+006mg/L @ 25°C (est). | |
C[NH2+]CCC[NH2+]C | |
104.20 | |
102.18 | |
N,N'-Dimethyl-1,3-propanediamine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 203-859-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only