Please login to your online account to display your discounted pricing

Alfa Aesar™ (?)-N,N-Dimethyl-1-[2-(diphenylphosphino)ferrocenyl]ethylamine monohydrate, 98%

$65.97 - $235.43

Chemical Identifiers

Chemical Name or Material (±)-N,N-Dimethyl-1-[2-(diphenylphosphino)ferrocenyl]ethylamine monohydrate
CAS 60816-98-0
Molecular Formula C26H28FeNP·H2O
Formula Weight 459.36g/mol (441.34g/mol Anhydrous)
MDL Number MFCD06199154
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1968403

 
alfa aesar™
L19684-03
1g Each for $235.43

AAL19684MD

 
alfa aesar™
L19684-MD
250mg Each for $65.97
Description & Specifications

Specifications

Melting Point 112° to 115°C
Quantity 1g
Chemical Name or Material (±)-N,N-Dimethyl-1-[2-(diphenylphosphino)ferrocenyl]ethylamine monohydrate
CAS 60816-98-0
Assay 98%
Molecular Formula C26H28FeNP·H2O
Formula Weight 459.36g/mol (441.34g/mol Anhydrous)
MDL Number MFCD06199154
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.