Learn More
N,N-Diisopropylethylamine trihydrofluoride, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 390710050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N, N-Diisopropylethylamine trihydrofluoride | |
94.0 | |
0.9650g/mL | |
40.5°C | |
90% | |
C8H22F3N | |
5g | |
diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-amine trihydrofluoride, n,n-diisopropylethylamine trihydrofluoride, acmc-1bx1e, ksc490i4d, n,n,-diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride, n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride, n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride, n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride | |
AJRRXKJZYYBJPY-UHFFFAOYSA-N | |
N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride | |
45108321 | |
90% |
131600-43-6 | |
100.0 | |
93°C to 95°C (4.0mmHg) | |
Authentic | |
Plastic bottle | |
MFCD00144880 | |
0.965 | |
Solubility in water: soluble | |
F.F.F.CCN(C(C)C)C(C)C | |
189.27 | |
189.26 |
Chemical Identifiers
131600-43-6 | |
189.27 | |
AJRRXKJZYYBJPY-UHFFFAOYSA-N | |
45108321 | |
F.F.F.CCN(C(C)C)C(C)C |
C8H22F3N | |
MFCD00144880 | |
diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-amine trihydrofluoride, n,n-diisopropylethylamine trihydrofluoride, acmc-1bx1e, ksc490i4d, n,n,-diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride, n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride, n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride, n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride | |
N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Rinse mouth.
Immediately call a POISON CENT
GHS Signal Word: Danger
RUO â Research Use Only