missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diisopropylethylamine 99.0+%, TCI America™

$28.80 - $166.75

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym 1,1'-dimethyltriethylamine, 2-propanamine, n-ethyl-n-1-methylethyl, diea, diisopropylethylamine, dipea, ethyldiisopropylamine, hunig's base, n,n-diisopropylethylamine, n-ethyl-n-isopropylpropan-2-amine, n-ethyldiisopropylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D159925ML
View Documents
Tci America
D159925ML
25mL Each for $28.80
 
D1599100ML
missing translation for 'viewDocuments'
Tci America
D1599100ML
100mL Each for $53.06
 
D1599500ML
missing translation for 'viewDocuments'
Tci America
D1599500ML
500mL Each for $166.75
 
Specifications

Chemical Identifiers

7087-68-5
129.247
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
1,1'-dimethyltriethylamine, 2-propanamine, n-ethyl-n-1-methylethyl, diea, diisopropylethylamine, dipea, ethyldiisopropylamine, hunig's base, n,n-diisopropylethylamine, n-ethyl-n-isopropylpropan-2-amine, n-ethyldiisopropylamine
N-ethyl-N-propan-2-ylpropan-2-amine

Specifications

127°C
-45°C
7087-68-5
MFCD00008868
1,1'-dimethyltriethylamine, 2-propanamine, n-ethyl-n-1-methylethyl, diea, diisopropylethylamine, dipea, ethyldiisopropylamine, hunig's base, n,n-diisopropylethylamine, n-ethyl-n-isopropylpropan-2-amine, n-ethyldiisopropylamine
CCN(C(C)C)C(C)C
129.247
129.25
≥99.0% (GC)
Yellow
25mL
C8H19N
2733
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
Clear Liquid at 20°C
N,N-Diisopropylethylamine
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3499

TSCA : Yes