Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diisopropylcarbodiimide, 99%

$95.23 - $162.73

Chemical Identifiers

Chemical Name or Material N,N'-Diisopropylcarbodiimide
CAS 693-13-0
Molecular Formula (CH3)2CHN=C=NCH(CH3)2
Formula Weight 126.2
MDL Number MFCD00065689
UN Number UN2929
Beilstein 878281
Synonym DIC
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1929209

  SDS
alfa aesar™
A1929209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1929214

  SDS
alfa aesar™
A1929214
25g Each for $95.23

AAA1929218

  SDS
alfa aesar™
A1929218
50g Each for $162.73

AAA1929230

  SDS
alfa aesar™
A1929230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.2
TSCA TSCA
CAS 693-13-0
Flash Point 33°C (91°F)
Beilstein 878281
UN Number UN2929
Quantity 10g
Odor Stench
MDL Number MFCD00065689
Chemical Name or Material N,N'-Diisopropylcarbodiimide
Health Hazard 2 GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 1 Danger
Boiling Point 145° to 148°C
Synonym DIC
RTECS Number FF2175000
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula (CH3)2CHN=C=NCH(CH3)2
Assay 99%
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.

Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.