Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisopropylbenzamide, 99%

$95.68 - $290.81

Chemical Identifiers

Chemical Name or Material N,N-Diisopropylbenzamide
CAS 20383-28-2
Molecular Formula C13H19NO
Formula Weight 205.3
MDL Number MFCD00026370
Beilstein 2048090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2291814

  SDS
alfa aesar™
B2291814
25g Each for $95.68

AAB2291822

  SDS
alfa aesar™
B2291822
100g Each for $290.81
Description & Specifications

Specifications

Formula Weight 205.3
Chemical Name or Material N,N-Diisopropylbenzamide
Melting Point 68° to 72°C
Boiling Point 148° to 152°C (18mmHg)
Assay 99%
Quantity 25g
Molecular Formula C13H19NO
MDL Number MFCD00026370
CAS 20383-28-2
Beilstein 2048090

Applications
N,N-Diisopropylbenzamide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 301 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.