Learn More
N,N-Diisopropylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5547106
Description
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.
Notes
Light Sensitive. Avoid oxidizing agents.
Specifications
N,N-Diisopropylaniline | |
Colorless to Yellow | |
95°C to 96°C (11 mmHg) | |
C12H19N | |
MFCD00048277 | |
Light sensitive | |
OVSARSKQWCLSJT-UHFFFAOYSA-N | |
N,N-di(propan-2-yl)aniline | |
61329 | |
97% |
4107-98-6 | |
0.91 g/mL | |
84°C (183°F) | |
1.519 | |
5 g | |
n,n-diisopropylaniline, benzenamine, n,n-bis 1-methylethyl, unii-kep9pka41k, kep9pka41k, n,n-dsopropylanlne, diisopropylphenyl amine, acmc-1asew, dsstox_cid_22189, dsstox_rid_79949, dsstox_gsid_42189 | |
CC(C)N(C1=CC=CC=C1)C(C)C | |
177.291 | |
177.29 | |
Liquid |
Chemical Identifiers
4107-98-6 | |
177.291 | |
OVSARSKQWCLSJT-UHFFFAOYSA-N | |
61329 | |
CC(C)N(C1=CC=CC=C1)C(C)C |
C12H19N | |
MFCD00048277 | |
n,n-diisopropylaniline, benzenamine, n,n-bis 1-methylethyl, unii-kep9pka41k, kep9pka41k, n,n-dsopropylanlne, diisopropylphenyl amine, acmc-1asew, dsstox_cid_22189, dsstox_rid_79949, dsstox_gsid_42189 | |
N,N-di(propan-2-yl)aniline |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only