missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisopropylaniline, 97%

Manufacturer:  Alfa Aesar™ H55471MD

 View more versions of this product

Catalog No. AAH55471MD



Description

Description

Applications
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

Notes
Light Sensitive. Avoid oxidizing agents.
Specifications

Specifications

N,N-Diisopropylaniline
1.5190
95°C to 96°C (11mmHg)
250mg
C12H19N
acmc-1asew
CC(C)N(C1=CC=CC=C1)C(C)C
177.291
177.29
97%
0.91g/mL
Light Sensitive
Colorless to Yellow
4107-98-6
MFCD00048277
OVSARSKQWCLSJT-UHFFFAOYSA-N
N,N-di(propan-2-yl)aniline
61329
Liquid

Chemical Identifiers

4107-98-6
177.291
OVSARSKQWCLSJT-UHFFFAOYSA-N
61329
CC(C)N(C1=CC=CC=C1)C(C)C
C12H19N
MFCD00048277
acmc-1asew
N,N-di(propan-2-yl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents