missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisopropyl-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 79606-48-7
Molecular Formula C13H18N2O3
Molecular Weight (g/mol) 250.298
MDL Number MFCD00584230
InChI Key RDBKRCFDYJOIDU-UHFFFAOYSA-N
Synonym n,n-diisopropyl-4-nitrobenzamide, 4-hydroxy oxido amino-n,n-diisopropylbenzamide, 4-nitro-n,n-di propan-2-yl benzamide, 4-nitro-n,n-bis propan-2-yl benzamide, n,n-diisopropyl-4-nitro-benzamide, n,n-bis methylethyl 4-nitrophenyl carboxamide
PubChem CID 347938
IUPAC Name 4-nitro-N,N-di(propan-2-yl)benzamide
SMILES CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60074MD View Documents Alfa Aesar™
H60074MD
250mg Each for $220.53
 
AAH6007403 View Documents Alfa Aesar™
H6007403
1g Each for $703.20
 
Specifications

Chemical Identifiers

79606-48-7
250.298
RDBKRCFDYJOIDU-UHFFFAOYSA-N
347938
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C13H18N2O3
MFCD00584230
n,n-diisopropyl-4-nitrobenzamide, 4-hydroxy oxido amino-n,n-diisopropylbenzamide, 4-nitro-n,n-di propan-2-yl benzamide, 4-nitro-n,n-bis propan-2-yl benzamide, n,n-diisopropyl-4-nitro-benzamide, n,n-bis methylethyl 4-nitrophenyl carboxamide
4-nitro-N,N-di(propan-2-yl)benzamide

Specifications

250mg
C13H18N2O3
5038710
RDBKRCFDYJOIDU-UHFFFAOYSA-N
4-nitro-N,N-di(propan-2-yl)benzamide
347938
97%
79606-48-7
MFCD00584230
n,n-diisopropyl-4-nitrobenzamide, 4-hydroxy oxido amino-n,n-diisopropylbenzamide, 4-nitro-n,n-di propan-2-yl benzamide, 4-nitro-n,n-bis propan-2-yl benzamide, n,n-diisopropyl-4-nitro-benzamide, n,n-bis methylethyl 4-nitrophenyl carboxamide
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
250.298
250.29
N,N-Diisopropyl-4-nitrobenzamide