missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisopropyl-3-nitrobenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 2451-01-6
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00594354
InChI Key JGKJMBOJWVAMIJ-UHFFFAOYSA-N
Synonym terpin hydrate, terpin hydrate, p-menthane-1,8-diol monohydrate, terpin monohydrate, terpin cis-form hydrate, 4-2-hydroxypropan-2-yl-1-methylcyclohexanol hydrate, terpin hydrate usp, terpin hydrate usp, terpin hydrate cis-form, cyclohexanemethanol, 4-hydroxy-alpha,alpha,4-trimethyl-, monohydrate
PubChem CID 17141
IUPAC Name 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate
SMILES CC1(CCC(CC1)C(C)(C)O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59903MD View Documents Alfa Aesar™
H59903MD
250mg Each for $211.27
 
AAH5990303 View Documents Alfa Aesar™
H5990303
1g Each for $675.45
 
Specifications

Chemical Identifiers

2451-01-6
190.28
JGKJMBOJWVAMIJ-UHFFFAOYSA-N
17141
CC1(CCC(CC1)C(C)(C)O)O.O
C10H22O3
MFCD00594354
terpin hydrate, terpin hydrate, p-menthane-1,8-diol monohydrate, terpin monohydrate, terpin cis-form hydrate, 4-2-hydroxypropan-2-yl-1-methylcyclohexanol hydrate, terpin hydrate usp, terpin hydrate usp, terpin hydrate cis-form, cyclohexanemethanol, 4-hydroxy-alpha,alpha,4-trimethyl-, monohydrate
4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate

Specifications

250mg
C13H18N2O3
n,n-diisopropyl-3-nitrobenzamide, 3-nitro-n,n-di propan-2-yl benzamide, acmc-209mhc, n,n-diisopropyl-3-nitro-benzamide, benzamide, n,n-diisopropyl-m-nitro, n,n-bis methylethyl 3-nitrophenyl carboxamide
CC(C)N(C(C)C)C(=O)C1=CC(=CC=C1)[N+](=O)[O-]
250.30
250.29
N,N-Diisopropyl-3-nitrobenzamide
2448-06-8
MFCD00594354
UFFKMMNKRCCXPE-UHFFFAOYSA-N
3-nitro-N,N-di(propan-2-yl)benzamide
221836
97%