missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisopropyl-3-methylbenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 5448-36-2
Molecular Formula C14H21NO
Molecular Weight (g/mol) 219.328
MDL Number MFCD00587389
InChI Key WPFBQNDWEOVSGC-UHFFFAOYSA-N
Synonym n,n-diisopropyl-3-methylbenzamide, 3-methyl-n,n-di propan-2-yl benzamide, 3-methyl-n,n-bis isopropyl benzamide, cbmicro_015287, cambridge id 5267036, n,n-bis methylethyl 3-methylphenyl carboxamide
PubChem CID 226948
IUPAC Name 3-methyl-N,N-di(propan-2-yl)benzamide
SMILES CC1=CC=CC(=C1)C(=O)N(C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59977MD View Documents Alfa Aesar™
H59977MD
250mg Each for $211.27
 
AAH5997703 View Documents Alfa Aesar™
H5997703
1g Each for $675.45
 
Specifications

Chemical Identifiers

5448-36-2
219.328
WPFBQNDWEOVSGC-UHFFFAOYSA-N
226948
CC1=CC=CC(=C1)C(=O)N(C(C)C)C(C)C
C14H21NO
MFCD00587389
n,n-diisopropyl-3-methylbenzamide, 3-methyl-n,n-di propan-2-yl benzamide, 3-methyl-n,n-bis isopropyl benzamide, cbmicro_015287, cambridge id 5267036, n,n-bis methylethyl 3-methylphenyl carboxamide
3-methyl-N,N-di(propan-2-yl)benzamide

Specifications

250mg
C14H21NO
n,n-diisopropyl-3-methylbenzamide, 3-methyl-n,n-di propan-2-yl benzamide, 3-methyl-n,n-bis isopropyl benzamide, cbmicro_015287, cambridge id 5267036, n,n-bis methylethyl 3-methylphenyl carboxamide
CC1=CC=CC(=C1)C(=O)N(C(C)C)C(C)C
219.328
219.33
N,N-Diisopropyl-3-methylbenzamide
5448-36-2
MFCD00587389
WPFBQNDWEOVSGC-UHFFFAOYSA-N
3-methyl-N,N-di(propan-2-yl)benzamide
226948
97%