missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diisobutyl-4-nitrobenzamide, 97%

Chemical Identifiers

CAS 139680-47-0
Molecular Formula C15H22N2O3
Molecular Weight (g/mol) 278.352
MDL Number MFCD01010614
InChI Key IFYJJQOERYQZBL-UHFFFAOYSA-N
Synonym n,n-bis 2-methylpropyl-4-nitrobenzamide, n,n-diisobutyl-4-nitrobenzamide, benzamide, n,n-bis 2-methylpropyl-4-nitro, n,n-bis 2-methylpropyl 4-nitrophenyl carboxamide
PubChem CID 347941
IUPAC Name N,N-bis(2-methylpropyl)-4-nitrobenzamide
SMILES CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60984MD View Documents Alfa Aesar™
H60984MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6098403 View Documents Alfa Aesar™
H6098403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

139680-47-0
278.352
IFYJJQOERYQZBL-UHFFFAOYSA-N
347941
CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C15H22N2O3
MFCD01010614
n,n-bis 2-methylpropyl-4-nitrobenzamide, n,n-diisobutyl-4-nitrobenzamide, benzamide, n,n-bis 2-methylpropyl-4-nitro, n,n-bis 2-methylpropyl 4-nitrophenyl carboxamide
N,N-bis(2-methylpropyl)-4-nitrobenzamide

Specifications

250mg
C15H22N2O3
n,n-bis 2-methylpropyl-4-nitrobenzamide, n,n-diisobutyl-4-nitrobenzamide, benzamide, n,n-bis 2-methylpropyl-4-nitro, n,n-bis 2-methylpropyl 4-nitrophenyl carboxamide
CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
278.352
278.35
N,N-Diisobutyl-4-nitrobenzamide
139680-47-0
MFCD01010614
IFYJJQOERYQZBL-UHFFFAOYSA-N
N,N-bis(2-methylpropyl)-4-nitrobenzamide
347941
97%