missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diisobutyl-1,6-hexanediamine, 97%

$62.22 - $230.38

Chemical Identifiers

CAS 16121-92-9
Molecular Formula (CH3)2CHCH2NH(CH2)6NHCH2CH(CH3)2
Formula Weight 228.41
MDL Number MFCD00048339
Chemical Name or Material N,N'-Diisobutyl-1,6-hexanediamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2102406 SDS Alfa Aesar™
B2102406
5g Each for $62.22
AAB2102414 SDS Alfa Aesar™
B2102414
25g Each for $230.38
Specifications Description & Specifications

Chemical Identifiers

16121-92-9
228.41
N,N'-Diisobutyl-1,6-hexanediamine
(CH3)2CHCH2NH(CH2)6NHCH2CH(CH3)2
MFCD00048339

Specifications

16121-92-9
(CH3)2CHCH2NH(CH2)6NHCH2CH(CH3)2
MFCD00048339
97%
5g
228.41
N,N'-Diisobutyl-1,6-hexanediamine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning